missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies MethoCult™ Express

Product Code. 17158221
Encompass
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17158221 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17158221 Supplier STEMCELL Technologies Supplier No. 04437

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Methylcellulose-based medium with recombinant cytokines for rapid hematopoietic colony assays of human cells

  • MethoCult™ Express is intended for use in hematopoietic colony-forming unit (CFU) assays of human cord blood (CB) samples, after a minimum culture period of 7 days
  • It is recommended for use with red blood cell-depleted CB samples, whole CB samples that have been cryopreserved and thawed, and CB mononuclear cells
  • MethoCult™ Express is optimized for the growth and enumeration of human hematopoietic progenitor cells, after a much shorter period than the 14 -16 days of standard CFU assays
  • In MethoCult™ Express, colonies containing at least 20 cells can be counted as early as after 7 days of culture
  • If MethoCult™ Express cultures are maintained for 14 - 16 days, colonies derived from erythroid progenitor cells (BFU-E), granulocyte-macrophage progenitor cells (CFU-GM, CFU-G, and CFU-M), and multi-potential granulocyte, erythroid, macrophage, and megakaryocyte progenitor cells (CFU-GEMM) can be counted

Specifications

Cell Type Hematopoietic Stem and Progenitor Cells
For Use With (Application) Cell Culture, Colony Assay, Functional Assay
Quantity 100 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.