missing translation for 'onlineSavingsMsg'
Learn More

Methanol, LC-MS Grade, Scharlab

Product Code. 11308434
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11308434 1 L 1 L
11318434 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11308434 Supplier Scharlab Supplier No. ME03261000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable for use as an eluent in LC-MS. Methanol, LC-MS Grade, Scharlab is free from alkaline impurities that can lead to the rise of ions that increase background noise and cause interference in the spectrum.

LC-MS is a powerful technique combining separation by HPLC with the structural information obtained by MS. Its application is widespread in the scientific and pharmaceutical fields as well as in biotechnology and pesticide residue analysis.

  • Designed with particularly low content (<100 ppb) of alkaline metals, especially K and Na
  • Microfiltered through a 0.22μm filter to eliminate any solid particles
  • Quality of the eluents is ensured by means of suitability tests for LC-MS
  • To improve ion generation, the addition of a volatile organic acid to the mobile phase is recommended

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Absorbance 0.699 AU at 205nm, 0.222 AU at 220nm, 0.046 AU at 240nm, 0.009 AU at 260nm
Color Colorless
Acidity Max. 0.0002meq/g
Alkalinity Max. 0.0002meq/g
Density 0.79g/cm3
Quantity 1 L
Flash Point 11°C
Identification Passes test
Ignition Point 455°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.3288
Residue after Evaporation Max. 0.0005%
Linear Formula CH3OH
Vapor Pressure 128 hPa
UN Number 1230
Water Max. 0.02%
Boiling Point 64.5°C
Solubility Information Miscible in water
Formula Weight 32.04g/mol
Percent Purity Min. 99.9%
Grade LC-MS
Chemical Name or Material Methanol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.