Learn More
Scharlab METANOL PURISS 5L
Brand:  Scharlab ME0301005P
METANOL PURISS 5L
Product Code. 11348434
  • ONLINE EXCLUSIVE PRICE
    137.55 EUR
    90.34 EUR / 5 L
    Save 47.21 EUR
    34% Off
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Methanol, extra pure, Pharmpur, Ph Eur, BP, NF
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301 + H311 + H331-Toxic if swallowed, in contact with skin or if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .