Learn More
Metamizol sodium monohydrate 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Product Code. 30314988
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30314988 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30314988 Supplier Dr. Ehrenstorfer Supplier No. DREA14942010AL1000
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Metamizole Sodium Monohydrate
CAS 5907-38-0
Analyte or Component Names Metamizole Sodium Monohydrate
Shipping Condition +20°C
Molecular Formula C13 H16 N3 O4 S . Na . H2 O
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Metamizole Sodium Monohydrate, 1-[(2,3-Dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)methylamino]-methanesulfonic acid monosodium salt monohydrate, (Antipyrinylmethylamino)-methanesulfonic acid sodium salt monohydrate, [(2,3-Dihydro-1,5-dimethyl-3-oxo-2-phenyl-1H-pyrazol-4-yl)methylamino]-methanesulfonic acid sodium salt monohydrate, Dipyrone monohydrate, Metamizol monohydrate, Sodium (antipyrinylmethylamino)methanesulfonate hydrate
InChI Formula InChI=1S/C13H17N3O4S.Na.H2O/c1-10-12(14(2)9-21(18,19)20)13(17)16(15(10)3)11-7-5-4-6-8-11;;/h4-8H,9H2,1-3H3,(H,18,19,20);;1H2/q;+1;/p-1
SMILES O.[Na+].CN(CS(=O)(=O)[O-])C1=C(C)N(C)N(C1=O)c2ccccc2
IUPAC Name sodium;[(1,5-dimethyl-3-oxo-2-phenylpyrazol-4-yl)-methylamino]methanesulfonate;hydrate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 351.35
Quantity 1 mL
Formula Weight 351.0865 g/mol