Learn More
Brady™ Metalized Polyester Labels for M6/M7 Printers
Click to view available options
Dimensions (LxW):
0.375 x 1 in.
0.5 x 1 in.
0.5 x 1.5 in.
0.75 x 1.5 in.
1 x 1.5 in.
1.5 x 1.5 in.
1.9 x 3 in.
1.9 x 4 in.
2 x 1 in.
For Use With (Equipment):
M610, M611, M710 (with Media Adapter), BMP61, BMP71 (with Media Adapter)
Height (Metric):
9.53 mm
12.7 mm
19.05 mm
25.4 mm
38.1 mm
48.26 mm
50.8 mm
Shape:
Rectangle
Square
Unit Size:
Pack of 100
Pack of 250
Pack of 500
Description
- Permanent acrylic adhesive withstands numerous solvents and variable temperatures
- UL approved, CSA accepted, RoHS Compliant
- Service temperature of -40°C to 90°C (-40°F to 194°F)
- Made of B-435A polyester material
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Certifications/Compliance | AGA Approved, ASTM D 1000, ASTM D 2979, ASTM G 155, CSA Approved, UL Recognized |
| Adhesive Type | Permanent Acrylic |
| Coating | None |
| Color | Silver |
| Finish | Gloss |
| Format | Die-cut |
| Material | Polyester |
| Printable Sides | Single-side Printable |
| Print Technology | Thermal Transfer |
| Show More |