Description
- Metal Detectable Volumetric Spoons are made from a special chemically resistant grade of PP.
- The whole range is manufactured in cleanroom conditions and packed in a class 7 (10,000) cleanroom.
- This ensures the cleanliness of the product.
- The robust design means they can be used for years and years or used as a single use product straight from the bag.
- Simply unwrap and use.
- They are ideal for use with powders, liquids and granules.
- The blue color means they can easily seen if left in the powder in line with HACCP guidelines.
- Made from high quality PP.
- Conforms to the latest FDA and EU food requirements.
- 2.5 mL volume is a standard size.
Specifications
Specifications
| Capacity (Metric) | 2.5 mL |
| Color | Blue |
| Material | Polypropylene |