Learn More
Sampling Systems™ Metal Detectable Powder Funnels
Product Code. 17554726 Shop All Sampling Systems Products
Change view
Click to view available options
Diameter (Metric)Top:
200 mm
Unit Size:
Pack of 4
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric)Top unitSize
17554726 200 mm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17554726 Supplier Sampling Systems™ Supplier No. M1051B200
Only null left
Add to basket
Add to basket
Our class leading X-ray and Metal Detectable Powder Funnels are available as standard in highly visible blue. The funnels are available with full food grade documentation.
  • The Sampling Systems metal detectable powder funnels are made from a special chemically resistant grade of PP.
  • The large spout means that powder does not clog and bridge when in use.
  • This means they are ideal for powders, creams and liquids.
  • The whole range is manufactured in cleanroom conditions and packed in an ISO class 7 (10,000) cleanroom.
  • This ensures the cleanliness of the product.
  • The robust design means they can be used for years and years or used as a single use product straight from the bag.
  • Simply unwrap and use.
  • They are ideal for use with liquids and suspensions.
  • Made from high quality PP.
  • Conforms to the latest FDA and EU food requirements.
  • The funnel has a large diameter spout to promote powder flow.
  • 200 mm diameter is a standard size.
Specifications
Diameter (Metric)Top 200 mm
Material Polypropylene
Color Blue