Description
- The Sampling Systems metal detectable powder funnels are made from a special chemically resistant grade of PP.
- The large spout means that powder does not clog and bridge when in use.
- This means they are ideal for powders, creams and liquids.
- The whole range is manufactured in cleanroom conditions and packed in an ISO class 7 (10,000) cleanroom.
- This ensures the cleanliness of the product.
- The robust design means they can be used for years and years or used as a single use product straight from the bag.
- Simply unwrap and use.
- They are ideal for use with liquids and suspensions.
- Made from high quality PP.
- Conforms to the latest FDA and EU food requirements.
- The funnel has a large diameter spout to promote powder flow.
- 200 mm diameter is a standard size.
Specifications
Specifications
| Diameter (Metric)Top | 200 mm |
| Material | Polypropylene |
| Color | Blue |