Description
- The detectable stirrer and pallet knives are made from a special chemically resistant grade of PP.
- The robust design means they can be used for years and years or used as a single use product straight from the bag.
- The whole range is manufactured in cleanroom conditions and packed in a class 7 (10,000) cleanroom.
- This ensures the purity and cleanliness of the product.
- Simply unwrap and use.
- Our ergonomic handle means they are easy to use.
- Many customers use these as stirrers.
- Made from high quality PP.
- Gives long lasting performance.
- Conforms to the latest FDA and EU food requirements.
- 200 mm blade is a standard size.
Specifications
Specifications
| Color | Blue |
| Width (Metric) Blade | 200 mm |
| Material | Polypropylene |