Learn More
DWK Life Sciences Metal Cap
123.50 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Use promo code "23258" to get your promotional price.
Click to view available options
Material:
Anodized Aluminum
Stainless Steel
Description
Microbiology is an important field of biology and medicine and deals with microorganisms (bacteria, fungi, protozoa or viruses) and their habitats and conditions. For this purpose, it is essential to produce pure culture media on which the corresponding microorganisms can grow. In order to avoid the introduction of foreign substances, the bottles and flasks must be covered but not sealed gas-tight. Air exchange is essential for culture growth.
- The metal caps allow an exchange of air in the media bottle, creating a positive environment for the growth of microorganisms
- These caps are autoclavable, but not dishwasher-safe
Specifications
Diameter (Metric) | 38 mm |
Color | Blue |
For Use With (Application) | Closuring of culture tubes or bottles; Closuring of culture media bottles; Storage and transport of Liquid cultures; Working with cell culture media under sterile conditions in biosafety cabinets and clean hoods |
Material | Anodized Aluminum |
Neck ID (Metric) | 38 mm |
Quantity | 10/Pk. |
Autoclavable | Autoclavable |
Fit for Purpose | This product is autoclavable, but it is not dishwasher-safe. |