missing translation for 'onlineSavingsMsg'
Learn More

Benzaldehyde in Acetonitrile 100μg/mL, Fisher Chemical™

Product Code. 12989593 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12989593 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12989593 Supplier Fisher Chemical Supplier No. P806180

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Specifications

Packaging Ampoule
CAS 1157-84-2
Physical Form Liquid
Molecular Formula C13H10N4O4
Concentration 100 μg/mL
MDL Number MFCD00156353
UN Number 1648
InChI Key DZPRPFUXOZTWAJ-NTEUORMPSA-N
Type Organic Single-component Standard Solution
SMILES [O-][N+](=O)C1=CC=C(N\N=C\C2=CC=CC=C2)C(=C1)[N+]([O-])=O
IUPAC Name (2E)-1-(2,4-dinitrophenyl)-2-(phenylmethylidene)hydrazine
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 286.25
PubChem CID 9566364
Quantity 1 mL
Chemical Name or Material Benzaldehyde
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Standard solution Benzaldehyde-DNPH 100ug/ml in acetonitrile
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P361 + P364-Take off immediately all contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to .
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.