Learn More
Meta-Phosphoric Acid, ACS Reagent, Honeywell Fluka™
Product Code. 15622440 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
6 x 500 g
Unit Size:
100g
2.5kg
500g
5g
Pack of 6
Product Code. 15622440
Brand: Honeywell-Fluka 239275500G
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 37267-86-0
Molecular Formula HO3P
Molecular Weight (g/mol) 79.979
MDL Number MFCD00011351
InChI Key UEZVMMHDMIWARA-UHFFFAOYSA-N
Synonym metaphosphoric acid, meta-phosphoric acid, hydrogenphosphonate, m-phosphoric acid, phosphenic acid, unii-mtk99r3uv0, metaphosphoric acid hpo3, x-phosphate-group, phosphonate 1-, hydrogen phosphonate
PubChem CID 3084658
SMILES OP(=O)=O
Specifications
Quantity 500 g
Linear Formula (HPO3)n
UN Number 3260
Grade ACS Reagent
Chemical Name or Material Meta-Phosphoric Acid