Learn More
[Met5, Lys6, Arg7] alpha-Neo-Endorphin (1-7), Thermo Scientific Chemicals
Product Code. 15497999
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
Product Code. 15497999
Brand: Thermo Scientific Alfa Aesar J66592.MA
Request Bulk or Custom Format
For cellular and molecular biology applications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 858.02
Product Type [Met5, Lys6, Arg7] alpha-Neo-Endorphin (1-7)
Quantity 10 mg
Regulatory Status RUO - research use only
Sequence Tyr-Gly-Gly-Phe-Met-Lys-Arg