Learn More
Mercury(Ii) Iodide Red, purum p.a, ≥99.0% (RT), Honeywell Fluka™
Product Code. 15607280 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 g
Packaging:
Glass bottle
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15607280 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15607280 Supplier Honeywell-Fluka Supplier No. 8337950G
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Puriss. p.a., ACS Reagent, red,≥99.0% (RT)
  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7774-29-0
Molecular Formula HgI2
Molecular Weight (g/mol) 454.401
MDL Number MFCD00011044
InChI Key YFDLHELOZYVNJE-UHFFFAOYSA-L
Synonym mercury diiodide, mercuric iodide, mercury ii iodide, red mercuric iodide, mercuric iodide, red, mercury biniodide, mercury iodide, mercurius bijodatus, hydrargyrum diodatum, hydrargyrum bijodatum
PubChem CID 24485
ChEBI CHEBI:49659
IUPAC Name diiodomercury
SMILES I[Hg]I
Specifications
Boiling Point 254°C
Melting Point 259°C
Color Red
Physical Form Powder
Quantity 50 g
UN Number UN1638
Formula Weight 454.4g/mol
Percent Purity ≥99.0% (RT)
Odor Odorless
Packaging Glass bottle
Chemical Name or Material Mercury(II) iodide red