Learn More
Emma Safety Footwear Mento Safety Shoes
Product Code. 15928771
Click to view available options
Size:
37
38
39
40
41
42
43
44
45
47
48
Unit Size:
1 pair
Product Code. 15928771
Brand: Emma Safety Footwear MM58154840
Water repellant for up to 4 hours and ideally suited to working outdoors. Designed to offer exceptional all over foot protection with a reinforced steel plate in the sole, and strong, supportive leather uppers. A double-density polyurethane sole protects feet from dangers above and below.
  • S3-Safety shoe with sole plate for wet work
  • Full grain leather construction
  • GripForce™ Easy Twist Outsole
  • Hydro-Tec™ Sanitized Silver provides drier, fresher feet
Specifications
Certifications/Compliance EN ISO 20345:2011
Color Brown
Design Mento
Liner Material Hydro-Tec™ Sanitized Silver
Material Leather
Size 40
Toe Material Steel