missing translation for 'onlineSavingsMsg'
Learn More

MELK-8a hydrochloride, MedChemExpress

Product Code. 16469536
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16469536

Brand: Medchem Express HY100368A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MELK-8a hydrochloride is a novel maternal embryonic leucine zipper kinase (MELK) inhibitor with an IC50 of 2 nM.

MELK-8a hydrochloride is a novel maternal embryonic leucine zipper kinase (MELK) inhibitor with an IC50 of 2 nM.

Specifications

Chemical Name or Material MELK-8a hydrochloride
Physical Form Solid
CAS 2096992-20-8
Color Light Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C25H33ClN6O
For Use With (Application) Cancer-Kinase/protease
Synonym NVS-MELK8a hydrochloride
Solubility Information H2O : ≥ 100 mg/mL (213.21 mM) ∣DMSO : 8.6 mg/mL (18.34 mM; Need ultrasonic and warming)
SMILES CN(CC1)CCN1C2=CC=C(N3C=C(C4=CC=NC=C4OCC5CCNCC5)C=N3)C=C2.Cl
Molecular Weight (g/mol) 469.02
Formula Weight 469.02
Percent Purity 99.26%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.