missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanoma Marker (MART-1 + gp100) Antibody (DT101+ BC199 + HMB45), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34608F
This item is not returnable.
View return policy
Description
Melanoma Marker (MART-1 + gp100) Monoclonal specifically detects Melanoma Marker (MART-1 + gp100) in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Melanoma Marker (MART-1 + gp100) | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| This antibody cocktail recognizes two melanoma-specific proteins, which include MART-1 and gp100. MART-1 is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Function of gp100 is not known but it is reported to be a useful marker for melanocytes and melanomas. A cocktail of these two markers is sensitive for labeling formalin-fixed, paraffin-embedded melanomas and other tumors showing melanocytic differentiation. | |
| Store at 4°C in the dark. |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| DT101+ BC199 + HMB45 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| MART1, MART-1 | |
| Recombinant hMART-1 protein (DT101& BC199); Extract of pigmented melanoma metastases from lymph nodes (HMB45) | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG2b κ, IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction