Learn More
VITLAB™ Melamine Formaldehyde Instrument Trays
Product Code. 15791996
Change view
Click to view available options
Dimensions (L x W x H):
190 x 150 x 40 mm
290 x 160 x 60 mm
350 x 250 x 40 mm
Unit Size:
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H) unitSize
15791996 290 x 160 x 60 mm Pack of 5
15781996 350 x 250 x 40 mm Pack of 5
15242619 190 x 150 x 40 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15791996 Supplier VITLAB™ Supplier No. 72398
Only null left
Add to basket
Add to basket
Use this practical tray for instruments, tools and sensitive utensils. VITLAB™ Melamine Formaldehyde Instrument Trays are stable and self-supporting with smooth, easy to clean surfaces.
  • White instrument tray
  • Tall shape
  • Rounded corners
  • Fitting lid made of PS, please order separately
Specifications
Dimensions (L x W x H) 290 x 160 x 60 mm
Color White
Height (Metric) 60 mm
Length (Metric) 290 mm
Material Melamine
Type Instrument Tray
Width (Metric) 160 mm
For Use With Practical tray for instruments, tools, and sensitive utensils