Learn More
OHAUS™ Medium Two Prong Sleeves
Product Code. p-7158987 Shop All Ohaus Products
Click to view available options
Material:
Fiberglass
Vinyl
Unit Size:
Each
Product Code. 15722669
Brand: OHAUS™ 30400253
Only null left
Add to basket
Add to basket
OHAUS™ Medium Two Prong Sleeves are for use with clamps and can be easily removed for cleaning or replacement.
  • Available in red vinyl and fiberglass
  • Fiberglass sleeves are ideal for application above 100°C
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2
Type Prong Sleeve
Material Vinyl
Size Medium
Color Red