missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Media Lab Bottle, Non-Graduated, PE Cap/Liner

Product Code. 12412511 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
125 mL
250 mL
500 mL
Quantity:
24 EA
48 EA
Unit Size:
Pack of 24
Pack of 48
This item is not returnable. View return policy

Product Code. 12412511

Brand: DWK Life Sciences 219495

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Clear Type I borosilicate glass bottle without graduations.

  • Manufactured from amber borosilicate glass which provides the best chemical resistance
  • Clear borosilicate glass bottle
  • Special pour lip facilitates pouring
  • Packaged in partitioned carton to help prevent scratch marks on bottles
  • Black Phenolic Polyethylene Lined Cap
  • Hazardous communication label included with each case of bottles allows for content ID and safety notification
  • Tamper evident seal included with each case of bottles keeps bottle sealed which protecting contents from accidental misuse
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 125 mL
Color Clear
Material Borosilicate Glass (Type I)
Quantity 48 EA
Closure Color Black
Neck Finish 33-340
Product Type Media Bottle
Closure Liner Polyethylene
Closure Material Phenolic
Closure Size 33-340 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.