Learn More
OHAUS™ Media Bottle Clamp
Product Code. 15792509 Shop All Ohaus Products
Click to view available options
For Use With (Equipment):
1000 mL Bottle
500 mL Bottle
Unit Size:
Each
Product Code. 15792509
Brand: OHAUS™ 30400097
Only null left
Add to basket
Add to basket
OHAUS™ Media Bottle Clamp is designed to securely grip and position various sizes of bottles.
  • Constructed of stainless steel
  • For use with media bottles
  • Designed to securely hold bottles in place
TRUSTED_SUSTAINABILITY
Specifications
Type Bottle Clamp
Material Stainless Steel
For Use With (Equipment) 500 mL Bottle
Description 500 mL Media Bottle Clamp