Learn More
MED8 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Product Code. 16819743 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16819743 20 μL 20µL
16809743 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16819743 Supplier Proteintech Supplier No. 121821AP20UL
Rabbit Polyclonal Antibody
This gene encodes a protein that is one of more than 20 subunits of the mediator complex, first identified in S. cerevisiae, that is required for activation of transcription. The product of this gene also interacts with elongins B and C, and CUL2 and RBX1, to reconstitute a ubiquitin ligase. Two alternative transcripts encoding different isoforms have been described.
Specifications
Antigen MED8
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.57 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MED8
Gene Accession No. Q96G25, Q9D7W5
Gene Alias ARC32, MED8, mediator complex subunit 8
Gene Symbols Med8
Host Species Rabbit
Immunogen MED8 Fusion Protein Ag2827
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112950, 80509
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG