missing translation for 'onlineSavingsMsg'
Learn More

MED6 Monoclonal Antibody (OTI3C9), TrueMAB™, OriGene

Product Code. 17559153
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17559153 30 μL 30µL
17539153 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17559153 Supplier Origene Technologies Supplier No. TA808183S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MED6 Monoclonal antibody specifically detects MED6 in Human samples. It is validated for Western Blot

Specifications

Antigen MED6
Applications Western Blot
Classification Monoclonal
Clone OTI3C9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene CDC42SE2
Gene Accession No. Q9NRR3
Gene Alias 1500012F11Rik; Activator-recruited cofactor 33 kDa component; ARC33; AU019660; AV213303; CTD-2540L5.5; hMed6; hypothetical protein LOC505293; MED6; Mediator complex subunit 6; mediator of RNA polymerase II transcriptio subunit 6-like protein; mediator of RNA polymerase II transcription subunit 6; mediator of RNA polymerase II transcription, subunit 6 homolog; NY-REN-28; Renal carcinoma antigen NY-REN-28
Host Species Mouse
Immunogen Full length human recombinant protein of human CDC42SE2 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56990
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.