missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED15 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
178.00 EUR - 383.00 EUR
Specifications
| Antigen | MED15 |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18377452
|
Novus Biologicals
NBP3-15565-20UL |
20 μg |
178.00 EUR
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18347184
|
Novus Biologicals
NBP3-15565-100UL |
100 μg |
383.00 EUR
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MED15 Polyclonal antibody specifically detects MED15 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| MED15 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS, 50% glycerol, pH7.3 | |
| 51586 | |
| IgG | |
| Affinity purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| ARC105Arc105, CAG7A, CTG repeat protein 7a, CTG7A, FLJ42282, FLJ42935, mediator complex subunit 15PC2 glutamine/Q-rich-associated protein, mediator of RNA polymerase II transcription subunit 15, PC2 (positive cofactor 2, multiprotein complex) glutamine/Q-rich-associatedprotein, PC2-glutamine-rich-associated protein, PCQAPDKFZp686A2214, Positive cofactor 2 glutamine/Q-rich-associated protein, positive cofactor 2, glutamine/Q-rich-associated protein, TIG1DKFZp762B1216, TIG-1TPA-inducible gene 1 protein, TNRC7, TPA inducible gene-1, TPA inducible protein, trinucleotide repeat containing 7,105-kD, Trinucleotide repeat-containing gene 7 protein | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MED15 (NP_001003891.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title