missing translation for 'onlineSavingsMsg'
Learn More

Meclozine Dihydrochloride, Mikromol™

Product Code. 17913516
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
missing translation for 'unitSize'
250mg
This item is not returnable. View return policy

Product Code. 17913516

missing translation for 'mfr': Mikromol MM0586.000250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Meclozine Dihydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Meclozine Hydrochloride
CAS 1104-22-9
Analyte or Component Names Meclozine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C25 H27 Cl N2 . 2 Cl H
Certifications/Compliance ISO 17034
Synonym Meclozine Dihydrochloride, 1-[(4-Chlorophenyl)phenylmethyl]-4-[(3-methylphenyl)methyl]piperazine dihydrochloride, Ancolan dihydrochloride, Bonamine, Diadril, Meclizine dihydrochloride, Meclizine hydrochloride, Meclozine dihydrochloride, Meclozine hydrochloride, NSC 28728, Navidoxine, Parachloramine dihydrochloride, Postafen, Postafene dihydrochloride, 1-(p-Chloro-alpha-phenylbenzyl)-4-(m-methylbenzyl)piperazine dihydrochloride
InChI Formula InChI=1S/C25H27ClN2.2ClH/c1-20-6-5-7-21(18-20)19-27-14-16-28(17-15-27)25(22-8-3-2-4-9-22)23-10-12-24(26)13-11-23;;/h2-13,18,25H,14-17,19H2,1H3;2*1H
SMILES Cl.Cl.Cc1cccc(CN2CCN(CC2)C(c3ccccc3)c4ccc(Cl)cc4)c1
IUPAC Name 1-[(4-chlorophenyl)-phenylmethyl]-4-[(3-methylphenyl)methyl]piperazine;dihydrochloride
Molecular Weight (g/mol) 463.87
Quantity 250 mg
Formula Weight 462.1396 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.