Description
- Mechanical Timer
- 60 min.
Specifications
Specifications
| Product Type | Mechanical Timer |
| Time Range | 60 min. |
| Dimensions (L x W x H) | 60 L x 60 H mm |
| Length (Metric) | 60 mm |
| Height (Metric) | 60 mm |
| Quantity | 1 Each |