missing translation for 'onlineSavingsMsg'
Learn More

Mebhydrolin, MedChemExpress

Product Code. 16448726
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448726 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16448726 Supplier Medchem Express Supplier No. HYB1303A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Mebhydrolin is a specific histamine H1 receptor antagonist.

Mebhydrolin is a specific histamine H1 receptor antagonist.

Specifications

Chemical Name or Material Mebhydrolin
CAS 524-81-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H20N2
Quantity 100 mg
Solubility Information DMSO : 50 mg/mL (180.91 mM; Need ultrasonic) ∣H2O : 1 mg/mL (3.62 mM; ultrasonic and warming and heat to 80°C)
SMILES CN(C1)CCC(N2CC3=CC=CC=C3)=C1C4=C2C=CC=C4
Molecular Weight (g/mol) 276.38
Formula Weight 276.38
Percent Purity 99.46%
Grade Research
Physical Form Powder
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.