Learn More
WTW™ Measuring Vessel for Electrodes
Click to view available options
For Use With (Application):
Determining pH of drinking water
Unit Size:
Each
Description
- Suitable for pH combination electrodes SenTix™ 81
- Also for use as a storage vessel for electrodes
- Supplied individually
Specifications
Specifications
| Type | Measuring Vessel |
| For Use With (Application) | Determining pH of drinking water |