missing translation for 'onlineSavingsMsg'
Learn More

MDK83190, MedChemExpress

Product Code. p-200100359
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16497548

Brand: Medchem Express HY18633

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MDK83190 is a potent apoptosis activator, induces Apaf-1 oligomerization, increases procaspase-9 processing and subsequent caspase-3 activation in a cyto c-dependent Manner.

MDK83190 is a potent apoptosis activator, induces Apaf-1 oligomerization, increases procaspase-9 processing and subsequent caspase-3 activation in a cyto c-dependent Manner.

Specifications

Chemical Name or Material MDK83190
Physical Form Solid
CAS 79183-19-0
Color Orange
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C15H9Cl2NO2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 46 mg/mL (150.26 mM)
SMILES O=C1N(CC2=CC=C(Cl)C(Cl)=C2)C3=C(C=CC=C3)C1=O
Molecular Weight (g/mol) 306.14
Formula Weight 306.14
Percent Purity 98.08%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.