missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDK Monoclonal Antibody (OTI6C8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807679S
This item is not returnable.
View return policy
Description
MDK Monoclonal antibody specifically detects MDK in Human samples. It is validated for Western BlotSpecifications
| MDK | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P48728 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 275 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI6C8 | |
| Unconjugated | |
| AMT | |
| amphiregulin-associated protein; ARAP; FLJ27379; MDK; Mek; Midgestation and kidney protein; midkine; midkine (neurite growth-promoting factor 2); MK; MK1; NEGF2; Neurite outgrowth-promoting factor 2; Neurite outgrowth-promoting protein; Retanoic acid-responsive protein; retinoic acid inducible factor; retinoic acid-induced differentiation factor; retinoic acid-responsive protein | |
| Human recombinant protein fragment corresponding to amino acids 146-403 of human AMT produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction