missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MD-2 Mouse anti-Human, Mouse, Rat, DyLight 680, Clone: 1A2E3, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-75513FR
514.25 EUR valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
MD-2 Monoclonal antibody specifically detects MD-2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| MD-2 | |
| Monoclonal | |
| DyLight 680 | |
| ESOP1, ESOP-1, Ly-96, lymphocyte antigen 96, MD2, MD-2, myeloid differentiation protein-2, Protein MD-2 | |
| Antibody was raised against a recombinant protein corresponding to amino acids 2 to 160 of human MD-2. | |
| 0.1 mL | |
| Signal Transduction | |
| 23643 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1A2E3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction