Learn More
VACUUBRAND™ MD 12C NT Chemistry Diaphragm Pump
GREENER_CHOICE
Product Code. 12643325
Click to view available options
Plug Type:
CEE
Unit Size:
Each
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 12643325
Brand: VACUUBRAND™ 20743300
Only null left
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Use this pump for continuous, oil-free pumping of corrosive gases and vapors and meet the highest requirements. Vacubrand™ MD 12C NT Chemistry Diaphragm Pumps provide the advantageous combination of high pumping speed and very low ultimate vacuum.
  • Outstanding chemical resistance and superior vapor tolerance
  • Reduced process time due to particularly high pumping speed even near ultimate vacuum
  • Whisper quiet and very low vibration
  • Very good ultimate vacuum even with open gas ballast valve for condensate purge
  • Long diaphragm lifetime, maintenance-free drive system
TRUSTED_SUSTAINABILITY
Specifications
Max. Vacuum 1.5 torr (without Gas Ballast), 3 torr (with Gas Ballast)
Horsepower 0.59 hp
Includes Manual
Motor Speed 150 to 1800 rpm
Certifications/Compliance ATEX Conformity II 3G IIC T3 X, CE, DIN EN 1012-2:2011, DIN EN 12100:2001, DIN EN 50581:2013, DIN EN 61010-1:2011, DIN EN 61326-1:2013, IEC 61010-1:2010
Electrical Requirements 230 V 50/60 Hz
Dimensions (L x W x H) 533 x 260 x 359 mm
Material Fluoroplastic (Case)
Model MD 12C NT
Noise Level 50 dbA
Plug Type CEE
Temperature (Metric) Storage -10°C to +60°C
Temperature (English) Max. 104°F
Temperature (Metric) Max 40°C
Temperature (Metric) Min. 10°C
Flow Rate 7.1/7.8 CFM
Frequency 50/60 Hz
Overall Height (Metric) 358.14 mm
Overall Length (Metric) 533.4 mm
Overall Width (Metric) 259.08 mm
Pump Type Diaphragm Pump
Enclosure Rating IP40
No. of Pump Heads 8
Resistance Properties Chemical-Resistant
Temperature (English) Min 50°F
Oil Type Oil-Free
No. of Stages 3
Maximum Pressure 16 psi
Voltage 230 V