Learn More
MCEE Monoclonal Antibody (OTI13B9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808575S
Encompass_Preferred
 View more versions of this product
Product Code. 17569983
  • 242.00 EUR / 30µL
Description
MCEE Monoclonal antibody specifically detects MCEE in Human samples. It is validated for Western Blot
Specifications
Specifications
MCEE
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q6UX39
Mouse
Affinity chromatography
RUO
401138
-20° C, Avoid Freeze/Thaw Cycles
IgG2a
Western Blot
OTI13B9
Unconjugated
AMTN
1110007A04Rik; DL-methylmalonyl-CoA racemase; GLOD2; glyoxalase domain containing 2; Mcee; methylmalonyl CoA epimerase; methylmalonyl-CoA epimerase; methylmalonyl-CoA epimerase, mitochondrial
Full length human recombinant protein of human AMTN produced in E.coli.
30 μL
Primary
Human
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions