Learn More
MCEE Monoclonal Antibody (OTI13B9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA808575
Encompass_Preferred
 View more versions of this product
Product Code. 17549983
  • 540.87 EUR / 100µL
Description
MCEE catalyzes the interconversion of D- and L-methylmalonyl-CoA during the degradation of branched chain amino acids. odd chain-length fatty acids, and other metabolites. Mutations in this gene result in methylmalonyl-CoA epimerase deficiency, which is presented as mild to moderate methylmalonic aciduria.
Specifications
Specifications
MCEE
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q96PE7
MCEE
Full length human recombit protein of human MCEE produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a
Western Blot
OTI13B9
Unconjugated
MCEE
GLOD2
Mouse
Affinity Chromatography
RUO
84693
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions