Learn More
MCEE Monoclonal Antibody (OTI13B9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808575
This item is not returnable.
View return policy
Description
MCEE catalyzes the interconversion of D- and L-methylmalonyl-CoA during the degradation of branched chain amino acids. odd chain-length fatty acids, and other metabolites. Mutations in this gene result in methylmalonyl-CoA epimerase deficiency, which is presented as mild to moderate methylmalonic aciduria.Specifications
| MCEE | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96PE7 | |
| MCEE | |
| Full length human recombit protein of human MCEE produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| OTI13B9 | |
| Unconjugated | |
| MCEE | |
| GLOD2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 84693 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |