missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBP Antibody, mFluor Violet 610 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24881MFV610
This item is not returnable.
View return policy
Description
MBP Polyclonal antibody specifically detects MBP in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MBP | |
| Polyclonal | |
| Western Blot | |
| MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
| Reacts with residues 49-62 [DRGAPKRGSGKDSH] of the 33 kDa human, mouse and rat MBP protein. Antibody detects primarily the 22 kDa isoform. | |
| 0.1 mL | |
| Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers | |
| 4155 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction