Learn More
Brannan™ Heavy Duty Max Min Thermometer
Click to view available options
Temperature Range (Metric):
-40°C to +50°C
Unit Size:
Each
Description
Clear and easy to read black temperature scale
- Wall mounting in plastic case
- 200mm long.
- Spirit filled, enclosed scale design with push button reset.
- Displays both °,C and °F
Specifications
Specifications
| Length (Metric) | 200 mm |
| Thermometer Fill | Red Spirit |
| Product Type | Analogue Thermometer |
| Max. Temperature (Metric) | 50°C |
| Min. Temperature (Metric) | -40°C |
| Temperature Range (Metric) | -40°C to +50°C |
| Accuracy | ±2.0°C |