missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Safety Products™ Maxi Welder Cut Gloves

Product Code. 12677513
Click to view available options
Size:
8
9
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12677513

Brand: Honeywell Safety Products™ 205859008

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Weld with superior protection using these MIG and arc welding, type B gloves. Honeywell Safety Products™ Maxi Welder Cut Gloves, made of cowhide and Kevlar™, are designed with flexible materials and features for excellent comfort.

  • Cowhide leather/Kevlar™ construction.
  • Winged thumb with yoke for flexibility and comfort.
  • Seamless forefinger.
  • Gauntlet type.
  • Silicone-coated Kevlar™ palm for heat and cut protection.
  • Kevlar™ fleece lining for heat insulation.
  • Inner lining of cotton for comfort and sweat absorption.
  • Water-repellent cowhide grain back.
  • Excellent resistance to contact and radiant heat.
  • Elastic on the back.
  • Kevlar™ stitching.
  • Split leather cuff to protect the forearm from mechanical hazards.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE CATEGORY II Intermediary risks, EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards, EN 407-04 Heat hazards, EN 12477-2000 Welding hazards, EC Category PPE 2, EC Certificate Number 0075/797/162/10/12/0807, EC Attestation Number 0075/797/162/10/12/0807
Classification Dexterity: 5
Coated Area Palm
Cuff Material Split Leather
Cut Style Gunn
Coating Material Silicone
Cut Resistance Rating 4
Material Grain Leather, Dupont™ Kevlar™ Aramid Fibers
Product Type Welding Gloves
Size 8
For Use With (Application) Automotive, Foundry, Iron, Steel, Ship Building, Transportation, Utility and Welding industries.
Glove Cuff Style Gauntlet
Liner Material Cotton
Lining Lined
Quantity 20/PK
Tear Rating 2
Length (Metric) 38.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.