missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Scie Plas Maxi-Plus Cooled Horizontal Gel Electrophoresis Unit
Click to view available options
Gel Dimensions:
300 x 250 mm
Unit Size:
Each
Description
- Robust acrylic tank construction 12mm walls, leak proof
- Safety lid with integral power lead
- Shrouded 4mm power output connectors universally compatible with modern-day low to medium voltage power supplies
- Long-life, 99.99% pure platinum electrodes
- Gold-plated terminals with corrosionfree stainless steel washers
- Durable high-impact polystyrene (HIPS) combs
- UV-transparent acrylic gel-casting trays
- Offset asymmetric electrodes
- Buffer recirculation ports
- Colour-coded combs corresponding to 1,1.5 and 2mm thicknesses
- Sde handle, for safe and easy transportation around baslaboratory
Specifications
Specifications
| Type | Horizontal Gel Unit |
| Gel Dimensions | 300 x 250 mm |
| Includes | Casting tray, 6 combs, buffer recirculation ports, connectors and loading strips |
| Model | CHU-25 |
| Sample Size | 624 |
| Dimensions (L x W) | 300 x 250 mm |
| Length (Metric) | 300 mm |
| Length (Metric) Gel | 300 mm |
| Width (Metric) Gel | 250 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction