missing translation for 'onlineSavingsMsg'
Learn More

IKA™ MATRIX PCR insert

Product Code. 16218701 Shop All IKA Products
Click to view available options
For Use With (Equipment):
IKA MATRIX Orbital
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 16218701

Brand: IKA™ 0030001159

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Insert for 0.2 ml PCR vials (included in scope of delivery of IKA MATRIX Orbital).

Suitable for: IKA MATRIX Orbital
TRUSTED_SUSTAINABILITY

Specifications

Holds PCR Reaction Vessels
Tube Capacity 0.2 mL
Product Type PCR Reaction Vessels, 96-Well PCR Plates for MATRIX Orbital
Dimensions (L x W x H) 128 x 86 x 30 mm
For Use With (Application) Samples with even the smallest volume, such as blood samples, pharmaceutical agents, ELISA assays/DNA/RNA samples
For Use With (Equipment) IKA MATRIX Orbital
No. of Positions 96
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.