Learn More
IKA™ MATRIX 1.5/2.0 ml insert
Product Code. 16208701 Shop All IKA Products
Click to view available options
For Use With (Equipment):
IKA MATRIX Orbital
Unit Size:
Each
Product Code. 16208701
Brand: IKA™ 0030001158
Only null left
Add to basket
Add to basket
Insert for 1.5/ 2.0 ml vials (included in scope of delivery of IKA MATRIX Orbital).
Suitable for: IKA MATRIX Orbital
TRUSTED_SUSTAINABILITY
Specifications
Holds Reaction Vessels
Tube Capacity 1.5/2 mL
Product Type Insert for Reaction Vessels for MATRIX Orbital
Dimensions (L x W x H) 128 x 86 x 45 mm
For Use With (Application) Samples with even the smallest volume, such as blood samples, pharmaceutical agents/DNA/RNA samples
For Use With (Equipment) IKA MATRIX Orbital
No. of Positions 24