missing translation for 'onlineSavingsMsg'
Learn More

MAT1A Rabbit anti-Human, Mouse, Plant, Rat, Polyclonal, Proteintech

Product Code. 16890094 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16890094 20 μL 20µL
16880094 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16890094 Supplier Proteintech Supplier No. 123951AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

This antibody was raised against the full length of human MAT1A and may cross-react with MAT2A. Western blot analysis using this antibody detected a single band around 44-50kD in liver.

MAT1A catalyzes the formation of S-adenosylmethionine from methionine and ATP. Methionine adenosyltransferase deficiency is caused by recessive and domit mutations, the latter identified in autosomal domit persistant hypermethioninemia.This gne encodes methionine adenosyltransferase I (alpha isoform), which catalyzes the formation of S-adenosylmethionine from methionine and ATP. Methionine adenosyltransferase deficiency is caused by recessive and domit mutations, the latter identified in autosomal domit persistant hypermethioninemia.

Specifications

Antigen MAT1A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.15 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene MAT1A
Gene Accession No. P13444, Q00266, Q91X83
Gene Alias AdoMet synthase 1, AMS1, MAT, MAT 1, MAT I/III, MAT1A, MATA1, SAMS, SAMS1
Gene Symbols MAT1A
Host Species Rabbit
Immunogen MAT1A Fusion Protein Ag3076
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11720, 25331, 4143
Target Species Rat, Mouse, Plant, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.