Learn More
Masterflex™ Pump Head Adapter
Description
- Cast aluminum adapters feature built-in gear box for easy setup
- Compatible with all I/P pump heads
- Suitable for use with 1⁄10 hp to 1⁄2 hp 56C frame motor and 650 rpm max pump head speed
- Flow rate 0.12 to 19 LPM with I/P tubing and 3.1:1 gear ratio—flow rate depends on drive rpm, pump head, and tubing size
Specifications
Specifications
| Flow Rate | 0.12 to 13 L/min. |
| Quantity | 1 Each |
| Type | Pump Head Adapter |