Learn More
Masterflex™ L/S™ High-Performance Precision Pump Tubing, Platinum-Cured Silicone
Product Code. p-8000837
Click to view available options
Tubing Size:
15
35
36
Unit Size:
7.6m
Each
Product Code. 11855181
Brand: Masterflex™ 9641015
Only null left
Add to basket
Add to basket
Excellent biocompatibility that meets multiple classifications
Ensure optimal performance from your Masterflex™ pump with this silicone pump tubing. Masterflex™ L/S™ Peroxide-Cured Silicone High-Performance Precision Pump Tubing provides accurate, repeatable flow. Ideal for pharmaceutical and biotechnology, peristaltic pumping and food applications.
  • Genuine Masterflex™ tubing undergoes robust testing to provide optimal performance/pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy, and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimize downtime
  • Regulatory Approvals to match your unique application
  • Always in stock — most formulations and sizes available to ship same day
  • Economical tubing with a longer tubing life
Specifications
Autoclavable Autoclavable
Material Platinum-cured Silicone
Diameter (English) Outer 0.59 in.
Diameter (Metric) Outer 15 mm
Overall Length (English) 24.9 ft.
Overall Length (Metric) 760 cm
For Use With (Application) Pharmaceutical, Biotechnology, Peristaltic Pumping and Food Applications
Min. Operating Temperature (Metric) -50°C
Tubing Size 15
Flow Rate 1.7 to 1000 mL/min
Hose Barb 3/16 in.
Height (Metric) 8.8 m
Color Translucent
Maximum Pressure 40 psi
Max. Operating Temperature (Metric) 230°C
Product Type Pump Tubing