Learn More
Masterflex™ Accessories for Peristaltic Pump Systems
Click to view available options
For Use With:
L/S digital drives
Unit Size:
Each
Description
Upgrade pumping and dispensing capabilities with Accessories for Masterflex™ Peristaltic Pump Systems.
- Accessory items for the Masterflex™ Peristaltic Pump Systems
Specifications
Specifications
| Description | Masterflex Momentary Start/stop Foot Switch |
| For Use With (Application) | Fluid handling |
| For Use With | L/S digital drives |
| Includes | DB25 connector, 1.8 m (6 ft.) cable |
| Quantity | 1 Ea. |
| Speed Control | Analog |
| Type | Foot Switch |