Learn More
Bolle Safety Masterall Safety Goggles
Description
- Equalizer technology filters and regulates moisture levels in goggles
- Permanent scratch-resistant platinum coating
- Sealed PVC frame and neoprene strap
- For use in offshore and petrochemical environments
Specifications
Specifications
| Lens Coating | Anti-fog, Scratch-resistant |
| Description | Double lens |
| Product Type | Safety goggles |
| For Use With (Application) | Offshore and petrochemical environments |
| Frame Type | Sealed PVC |