Learn More
Atago™ MASTER-BR Model Coolant Refractometer
Click to view available options
Unit Size:
Each
Specifications
Specifications
| Type | Coolant Refractometer |
| Scale Range | 0 to 70% Ethylene Glycol, 0°C to -50°C, 0 to 70% Propylene Glycol, 0°C to -50°C |
| Model | MASTER-BR |
| Scale Type | E.G/P.G |
| For Use With (Application) | Measuriing the concentration and freezing temperature of ethylene glycol and propylene glycol |
| Weight (Metric) | 90 g |