missing translation for 'onlineSavingsMsg'
Learn More

MASP2 Monoclonal Antibody (OTI7D4), TrueMAB™, OriGene

Product Code. 17507014
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17507014 30 μL 30µL
17587004 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17507014 Supplier Origene Technologies Supplier No. TA812532S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

MASP2 Monoclonal antibody specifically detects MASP2 in Human samples. It is validated for Western Blot

Specifications

Antigen MASP2
Applications Western Blot
Classification Monoclonal
Clone OTI7D4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene MASP2
Gene Accession No. O00187
Gene Alias mannan binding lectin serine peptidase 2; mannan-binding; mannan-binding lectin serine peptidase 1 pseudogene 1; mannan-binding lectin serine peptidase 2; mannan-binding lectin serine peptidase 2 variant 1; mannan-binding lectin serine peptidase 2 variant 2; mannan-binding lectin serine peptidase 2 variant 3; mannan-binding lectin serine peptidase 2 variant 4; mannan-binding lectin serine protease 1 pseudogene 1; mannan-binding lectin serine protease 2; Mannan-binding lectin serine protease 2 A chain; Mannan-binding lectin serine protease 2 B chain; mannose binding lectin-associated serine protease-2; mannose-binding protein-associated serine protease 2; MAp19; MASP1P1; Masp2; MASP-2; MBL-associated plasma protein of 19 kD; MBL-associated serine protease 2; sb:eu714; small MBL-associated protein; sMAP; wu:fb62a09
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 445-686 of human MASP2 (NP_006601) produced in E.coli.
Purification Method Protein A/G
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10747
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.