missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Masitinib mesylate, MedChemExpress
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
Description
Masitinib mesylate (AB-1010 mesylate) is a potent, orally bioavailable, and selective inhibitor of c-Kit (IC50=200 nM for human recombinant c-Kit). It also inhibits PDGFRα/β (IC50s=540/800 nM), Lyn (IC50= 510 nM for LynB), Lck, and, to a lesser extent, FGFR3 and FAK. Masitinib mesylate (AB-1010 mesylate) has anti-proliferative, pro-apoptotic activity and low toxicity.
Specifications
Specifications
| Chemical Name or Material | Masitinib mesylate |
| CAS | 1048007-93-7 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C29H34N6O4S2 |
| Quantity | 10 mg |
| Synonym | AB-1010 mesylate |
| Solubility Information | DMSO : ≥ 30 mg/mL (50.44 mM) |
| SMILES | O=C(NC1=CC=C(C(NC2=NC(C3=CC=CN=C3)=CS2)=C1)C)C4=CC=C(CN5CCN(CC5)C)C=C4.CS(=O)(O)=O |
| Molecular Weight (g/mol) | 594.75 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction