Learn More
Brady™ Marine Labels for BradyJet J4000 and J5000, BS 5609
Product Code. 17449202
Click to view available options
Height (Metric):
85 mm
110 mm
155 mm
210 mm
279.4 mm
Width (Metric):
55 mm
75 mm
105 mm
154.9 mm
203 mm
Unit Size:
Each
Product Code. 17449202
Brand: Brady™ 150756
Only null left
Add to basket
Add to basket
Don’t let a label on the final product sink your efficiency. With BS 5609 Marine Labels and the BradyJet J4000 or J5000 Printer, your labels stay stuck, even at sea.
  • Keep it colorful- this material runs through the BradyJet J4000 or J5000 Inkjet printer for high-resolution, full-color printing
  • An ink cartridge is required to print this material
Specifications
Autoclavable Not Autoclavable
Certifications/Compliance BS 5609:1986 Section 2 and section 3, Compliant with Directive 2011/65/EU and 2015/863
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Die-cut
Material Polypropylene (B-2585)
Printable Sides Single-side Printable
Print Technology Ink Jet
Product Type Label
Sterility Non-sterile
For Use With (Application) Hazard Communication Identification, Marine Labeling
For Use With (Equipment) BradyJet J4000 and J5000 Color Label Printers
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
No. per Pack 120 Labels/Roll
Quantity 120 Labels
Shape Rectangle
Temperature Range (English) -40°F to +140°F
Temperature Range (Metric) -40°C to +60°C
Thickness (English) 0.0088 in.
Thickness (Metric) 0.22 mm
Dimensions (LxW) 210.82 x 154.94 mm
Length (English) 8.3 in.
Width (English) 6.1 in.
Width (Metric) 154.9 mm
Height (English) 8.3 in.
Height (Metric) 210 mm
Alternate Part Number J50-267-2585