Learn More
SCAT™ Safety Funnel 'MARCO' with Lid, V2.0
Product Code. p-7203080 Shop All SCAT Europe Products
missing translation for 'orderingAttributeHoverText'
Height (Metric):
150 mm
370 mm
125 cm
Includes:
Removable Sieve, Earthing Cable
Removable Sieve, Earthing Cable, 220 mm Lance
Removable Sieve, Earthing Cable, Level Control w/ 150 mm Floater
Removable Sieve, Earthing Cable, Level Control w/ 190 mm Floater
Thread Size (Metric):
GL45
R2 in. BSP/G2 (m), 2 in. Tri-Sure (m)
S50
S51
S55
S60/61
S65
S70/71
missing translation for 'unitSize'
Each
Product Code. 16852130
missing translation for 'mfr': SCAT™ 318962
Only null left
Add to basket
Add to basket
Can be safely closed to prevent danger of ignition.
  • Modern lid funnel made of high-quality HDPE with very high resistance to acids, alkalis and many other chemicals
  • Integrated gripping edge on funnel lid increases grip without having to touch possibly contoured lid
  • Innovative funnel lid with integrated sealing ring closes funnel edge to prevent escape of explosive or health-endangering gases
  • Sturdy hinge guarantees long service life of the closing mechanism
  • Rotating screw cap makes it easier to screw lid funnel on and off the canister
  • Removable sieve for easy cleaning
  • Conductive version of MARCO V2.0 is black, differring in color from the non-conductive version
Specifications
Diameter (Metric)Top 140 mm
Capacity (Metric) 300 mL
Material HDPE-EC
Height (Metric) 150 mm
Autoclavable Not Autoclavable
Color Black
Includes Removable Sieve, Earthing Cable
Thread Size (Metric) GL45
Product Type Safety Funnel with Hinged Lid