Learn More
Maprotiline hydrochloride, MedChemExpress
Product Code. 16428316
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 16428316
Brand: Medchem Express HYB0444
MedChemExpress Maprotiline hydrochloride is a selective noradrenalin re-uptake inhibitor and a tetracyclic antidepressant.
Maprotiline hydrochloride is a selective noradrenalin re-uptake inhibitor and a tetracyclic antidepressant.
Specifications
Chemical Name or Material Maprotiline hydrochloride
CAS 10347-81-6
Color White
Purity Grade Notes Research
Molecular Formula C20H24ClN
Quantity 100 mg
Solubility Information DMSO : 83.33 mg/mL (265.50 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 2.2 mg/mL (7.01 mM; Need ultrasonic)
SMILES CNCCCC1(CC2)C3=CC=CC=C3C2C4=CC=CC=C14.Cl
Molecular Weight (g/mol) 313.86
Formula Weight 313.86
Percent Purity 99.43%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation